Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Bromomethyl)-4-fluoronaphthalene 98%

Supplier: AMBEED, INC
Danger

A237819-100MG A237819-1G A237819-250MG A237819-5G
77140-192EA 32.96 USD InStock
77140-192 77140-194 77140-196 77140-198
1-(Bromomethyl)-4-fluoronaphthalene 98%
1-(Bromomethyl)-4-fluoronaphthalene
Precaución: for Research Use Only


Formula: C₁₁H₈BrF
MDL Number: MFCD12025245
CAS Number: 6905-05-1
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty