Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-S-tert-Butyl 2-methylpropane-2-sulfinothioate 95%

Supplier: AMBEED, INC
Warning

A240301-100MG A240301-1G A240301-250MG A240301-5G
77140-230EA 53.56 USD InStock
77140-230 77140-232 77140-234 77140-236
(R)-S-tert-Butyl 2-methylpropane-2-sulfinothioate 95%
(R)-S-tert-Butyl 2-methylpropane-2-sulfinothioate
Precaución: for Research Use Only


Formula: C₈H₁₈OS₂
MW: 194.358 g/mol
MDL Number: MFCD08460193
CAS Number: 67734-35-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty