Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dichlorobis[2-(diphenylphosphino)ethylamine]ruthenium(II) 98%

Supplier: AMBEED, INC
Warning

A241310-100MG A241310-1G A241310-250MG
77140-270EA 62.83 USD InStock
77140-270 77140-272 77140-274
Dichlorobis[2-(diphenylphosphino)ethylamine]ruthenium(II) 98%
Dichlorobis[2-(diphenylphosphino)ethylamine]ruthenium(II)
Precaución: for Research Use Only


Formula: C₂₈H₃₂Cl₂N₂P₂Ru
MW: 630.4906 g/mol
MDL Number: MFCD07782004
CAS Number: 506417-41-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty