Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromobenzoxazole 97%

Supplier: AMBEED, INC
Warning

A242178-100G A242178-10G A242178-1G A242178-250MG A242178-25G A242178-5G
77140-298EA 593.28 USD InStock
77140-298 77140-300 77140-302 77140-304 77140-306 77140-308
5-Bromobenzoxazole 97%
5-Bromobenzoxazole
Precaución: for Research Use Only


Formula: C₇H₄BrNO
MDL Number: MFCD03095026
CAS Number: 132244-31-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty