Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-1,3'-Bipyrrolidine 98%

Supplier: AMBEED, INC
Warning

A246893-100MG A246893-1G A246893-250MG
77140-402EA 43.25 USD InStock
77140-402 77140-404 77140-406
(R)-1,3'-Bipyrrolidine 98%
(R)-1,3'-Bipyrrolidine
Precaución: for Research Use Only


Formula: C₈H₁₆N₂
MDL Number: MFCD11519103
CAS Number: 900164-95-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty