Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluoro-4-nitroanisole 97%

Supplier: AMBEED, INC
Warning

A247607-100G A247607-10G A247607-1G A247607-25G A247607-5G
77140-426EA 271.92 USD InStock
77140-426 77140-428 77140-430 77140-432 77140-434 77140-436
2,3-Difluoro-4-nitroanisole 97%
2,3-Difluoro-4-nitroanisole
Precaución: for Research Use Only


Formula: C₇H₅F₂NO₃
MDL Number: MFCD07995055
CAS Number: 66684-59-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty