Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Mdivi -1 98%

Supplier: AMBEED, INC
Warning

A252480-10MG A252480-1MG A252480-5MG A252480-1G
77140-512EA 32.96 USD InStock
77140-512 77140-514 77140-516 77685-717
Mdivi -1 98%
Mdivi -1
Precaución: for Research Use Only


Formula: C₁₅H₁₀Cl₂N₂O₂S
MDL Number: MFCD00974506
CAS Number: 338967-87-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty