Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Amino-PEG1-acid 97%

Supplier: AMBEED, INC
Warning

A253281-100MG A253281-1G A253281-250MG A253281-5G A253281-25G
77140-548EA 45.33 USD InStock
77140-548 77140-550 77140-552 77140-554 77746-406
Amino-PEG1-acid 97%
Amino-PEG1-acid
Precaución: for Research Use Only


Formula: C₅H₁₁NO₃
MDL Number: MFCD22017475
CAS Number: 144942-89-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty