Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(3-(4-((1H-Indazol-5-yl)amino)quinazolin-2-yl)phenoxy)-N-isopropylacetamide 99%

Supplier: AMBEED, INC
Warning

A255643-10MG A255643-1MG A255643-25MG A255643-5MG
77140-622EA 9.27 USD InStock
77140-622 77140-624 77140-626 77140-628 77140-630
2-(3-(4-((1H-Indazol-5-yl)amino)quinazolin-2-yl)phenoxy)-N-isopropylacetamide 99%
2-(3-(4-((1H-Indazol-5-yl)amino)quinazolin-2-yl)phenoxy)-N-isopropylacetamide
Precaución: for Research Use Only


Formula: C₂₆H₂₄N₆O₂
MDL Number: MFCD23098791
CAS Number: 911417-87-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty