Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tildipirosin 98%

Supplier: AMBEED, INC
Warning

A262611-10MG A262611-1MG A262611-25MG A262611-50MG A262611-5MG A262611-1G A262611-250MG A262611-5G
77140-756EA 8.24 USD InStock
77140-756 77140-758 77140-760 77140-762 77140-764 77685-748 77685-749 77685-750
Tildipirosin 98%
Tildipirosin
Precaución: for Research Use Only


Formula: C₄₁H₇₁N₃O₈
MDL Number: MFCD13194925
CAS Number: 328898-40-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty