Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ammonium decanedioate 98%

Supplier: AMBEED, INC
Warning

A266834-100G A266834-1G A266834-25G
77140-906EA 11.33 USD InStock
77140-906 77140-908 77140-910
Ammonium decanedioate 98%
Ammonium decanedioate
Precaución: for Research Use Only


Formula: C₁₀H₂₄N₂O₄
MW: 236.31 g/mol
MDL Number: MFCD00067242
CAS Number: 19402-63-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty