Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-fluorobenzyl alcohol 97%

Supplier: AMBEED, INC
Warning

A273763-100G A273763-1G A273763-250MG A273763-25G A273763-5G
77141-012EA 213.21 USD InStock
77141-012 77141-014 77141-016 77141-018 77141-020
4-Bromo-3-fluorobenzyl alcohol 97%
4-Bromo-3-fluorobenzyl alcohol
Precaución: for Research Use Only


Formula: C₇H₆BrFO
MDL Number: MFCD08236860
CAS Number: 222978-01-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty