Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-5-methoxy-2-(trifluoromethyl)quinazoline 97%

Supplier: AMBEED, INC
Warning

A275535-100MG A275535-1G
77141-036EA 488.24 USD InStock
77141-036 77141-038
4-Chloro-5-methoxy-2-(trifluoromethyl)quinazoline 97%
4-Chloro-5-methoxy-2-(trifluoromethyl)quinazoline
Precaución: for Research Use Only


Formula: C₁₀H₆ClF₃N₂O
MW: 262.62 g/mol
MDL Number: MFCD29923018
CAS Number: 219773-96-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty