Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1S)-(-)-Camphanic chloride 95%

Supplier: AMBEED, INC
Danger

A276987-1G A276987-250MG A276987-25G A276987-5G
77141-082EA 16.48 USD InStock
77141-082 77141-084 77141-086 77141-088
(1S)-(-)-Camphanic chloride 95%
(1S)-(-)-Camphanic chloride
Precaución: for Research Use Only


Formula: C₁₀H₁₃ClO₃
MW: 216.67 g/mol
MDL Number: MFCD00135626
CAS Number: 39637-74-6
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty