Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2,2-Dimethoxyethyl)-5-methoxy-6-(methoxycarbonyl)-4-oxo-1,4-dihydropyridine-3-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A277021-100G A277021-10G A277021-250MG A277021-25G A277021-5G A277021-1G A277021-500G
77141-090EA 233.81 USD InStock
77141-090 77141-092 77141-094 77141-096 77141-098 77746-495 77746-496
1-(2,2-Dimethoxyethyl)-5-methoxy-6-(methoxycarbonyl)-4-oxo-1,4-dihydropyridine-3-carboxylic acid 98%
1-(2,2-Dimethoxyethyl)-5-methoxy-6-(methoxycarbonyl)-4-oxo-1,4-dihydropyridine-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₇NO₈
MDL Number: MFCD28978322
CAS Number: 1335210-23-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty