Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Benzoylpyrrole 98%

Supplier: AMBEED, INC
Warning

A285601-25G A285601-5G A285601-1G
77141-240EA 97.85 USD InStock
77141-240 77141-242 77685-811
2-Benzoylpyrrole 98%
2-Benzoylpyrrole
Precaución: for Research Use Only


Formula: C₁₁H₉NO
MDL Number: MFCD00094048
CAS Number: 7697-46-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty