Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-4-fluorobenzoic acid 98%

Supplier: AMBEED, INC
Warning

A287952-10G A287952-25G A287952-5G A287952-100G
77141-288EA 11.33 USD InStock
77141-288 77141-290 77141-292 77685-822
3-Chloro-4-fluorobenzoic acid 98%
3-Chloro-4-fluorobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₄ClFO₂
MW: 174.56 g/mol
Boiling Pt: 273
Melting Pt: 136
MDL Number: MFCD00042477
CAS Number: 403-16-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty