Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl fumarate 98%

Supplier: AMBEED, INC
Warning

A288473-100G A288473-25G A288473-500G
77141-294EA 15.45 USD InStock
77141-294 77141-296 77141-298
Diethyl fumarate 98%
Diethyl fumarate
Precaución: for Research Use Only


Formula: C₈H₁₂O₄
MW: 172.18 g/mol
Boiling Pt: 219
Flash Pt: 91
MDL Number: MFCD00064455
CAS Number: 623-91-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty