Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-Fluorophenyl)glycine 98%

Supplier: AMBEED, INC
Warning

A291892-1G A291892-250MG A291892-5G
77141-346EA 130.81 USD InStock
77141-346 77141-348 77141-350
N-(4-Fluorophenyl)glycine 98%
N-(4-Fluorophenyl)glycine
Precaución: for Research Use Only


Formula: C₈H₈FNO₂
MDL Number: MFCD02255629
CAS Number: 351-95-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty