Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-((2R,4S,5R)-4-((tert-Butyldimethylsilyl)oxy)-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione 97%

Supplier: AMBEED, INC
Warning

A294505-100MG A294505-1MG A294505-250MG A294505-25G
77141-398EA 45.32 USD InStock
77141-398 77141-400 77141-402 77746-550
1-((2R,4S,5R)-4-((tert-Butyldimethylsilyl)oxy)-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione 97%
1-((2R,4S,5R)-4-((tert-Butyldimethylsilyl)oxy)-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione
Precaución: for Research Use Only


Formula: C₁₆H₂₈N₂O₅Si
MW: 356.48 g/mol
MDL Number: MFCD01631040
CAS Number: 40733-27-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty