Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-bromobenzoate 97%

Supplier: AMBEED, INC
Warning

A296997-100G A296997-10G A296997-1G A296997-25G A296997-500G A296997-5G
77141-430EA 41.2 USD InStock
77141-430 77141-432 77141-434 77141-436 77141-438 77141-440
Methyl-3-bromobenzoate 97%
Methyl-3-bromobenzoate
Precaución: for Research Use Only


Formula: C₈H₇BrO₂
MDL Number: MFCD00017777
CAS Number: 618-89-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty