Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Allylaminocarbonyl)benzeneboronic acid 96%

Supplier: AMBEED, INC
Warning

A310150-100MG A310150-1G A310150-250MG A310150-5G
77141-654EA 6.18 USD InStock
77141-654 77141-656 77141-658 77141-660
3-(Allylaminocarbonyl)benzeneboronic acid 96%
3-(Allylaminocarbonyl)benzeneboronic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₂BNO₃
MDL Number: MFCD04115702
CAS Number: 850567-29-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty