Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thieno[2,3-b]pyridine-2-carboxaldehyde 97%

Supplier: AMBEED, INC
Warning

A310173-100MG A310173-1G A310173-250MG
77141-662EA 33.99 USD InStock
77141-662 77141-664 77141-666
Thieno[2,3-b]pyridine-2-carboxaldehyde 97%
Thieno[2,3-b]pyridine-2-carboxaldehyde
Precaución: for Research Use Only


Formula: C₈H₅NOS
MDL Number: MFCD07772791
CAS Number: 53174-98-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty