Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl (6-(hydroxymethyl)pyridin-3-yl)carbamate 95%

Supplier: AMBEED, INC
Warning

A311129-100MG A311129-1G A311129-250MG
77141-684EA 110.2 USD InStock
77141-684 77141-686 77141-688
tert-Butyl (6-(hydroxymethyl)pyridin-3-yl)carbamate 95%
tert-Butyl (6-(hydroxymethyl)pyridin-3-yl)carbamate
Precaución: for Research Use Only


Formula: C₁₁H₁₆N₂O₃
MW: 224.26 g/mol
MDL Number: MFCD12031249
CAS Number: 323578-38-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty