Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Pyridylacetonitrile hydrochloride 98%

Supplier: AMBEED, INC
Warning

A313232-100MG A313232-1G A313232-250MG
77141-706EA 11.33 USD InStock
77141-706 77141-708 77141-710
4-Pyridylacetonitrile hydrochloride 98%
4-Pyridylacetonitrile hydrochloride
Precaución: for Research Use Only


Formula: C₇H₇ClN₂
MDL Number: MFCD00012825
CAS Number: 92333-25-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty