Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-6-(trifluoromethyl)benzaldehyde 95%

Supplier: AMBEED, INC
Warning

A313761-100MG A313761-250MG A313761-5G
77141-716EA 69.01 USD InStock
77141-716 77141-718 77685-892
2-Bromo-6-(trifluoromethyl)benzaldehyde 95%
2-Bromo-6-(trifluoromethyl)benzaldehyde
Precaución: for Research Use Only


Formula: C₈H₄BrF₃O
MDL Number: MFCD22381476
CAS Number: 1236141-95-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty