Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Fluorophenyl)(4-fluorophenyl)(phenyl)methanol 98+%

Supplier: AMBEED, INC
Warning

A317002-1G A317002-250MG A317002-5G
77141-744EA 16.48 USD InStock
77141-744 77141-746 77141-748
(2-Fluorophenyl)(4-fluorophenyl)(phenyl)methanol 98+%
(2-Fluorophenyl)(4-fluorophenyl)(phenyl)methanol
Precaución: for Research Use Only


Formula: C₁₉H₁₄F₂O
MDL Number: MFCD25977181
CAS Number: 128092-72-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty