Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate 98%

Supplier: AMBEED, INC
Warning

A330881-100MG A330881-10G A330881-1G A330881-250MG A330881-5G
77141-952EA 49.44 USD InStock
77141-952 77141-954 77141-956 77141-958 77141-960
1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate 98%
1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate
Precaución: for Research Use Only


Formula: C₃₄H₃₈F₆IrP₃
MDL Number: MFCD00064800
CAS Number: 38465-86-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty