Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Cyano-3,5-difluorophenylboronic acid 95%

Supplier: AMBEED, INC
Warning

A332381-100MG A332381-250MG
77141-962EA 18.54 USD InStock
77141-962 77141-964
4-Cyano-3,5-difluorophenylboronic acid 95%
4-Cyano-3,5-difluorophenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₄BF₂NO₂
MW: 182.92 g/mol
MDL Number: MFCD07644470
CAS Number: 861710-03-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty