Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(tert-Butyl)-4-chloroquinazoline 97%

Supplier: AMBEED, INC
Warning

A334675-100MG A334675-1G A334675-250MG
77141-982EA 12.36 USD InStock
77141-982 77141-984 77141-986
2-(tert-Butyl)-4-chloroquinazoline 97%
2-(tert-Butyl)-4-chloroquinazoline
Precaución: for Research Use Only


Formula: C₁₂H₁₃ClN₂
MW: 220.69 g/mol
MDL Number: MFCD20257998
CAS Number: 403612-89-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty