Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Ethoxy-2-fluoro-5-methylphenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A342895-1G A342895-250MG A342895-5G
77142-112EA 143.17 USD InStock
77142-112 77142-114 77142-116
3-Ethoxy-2-fluoro-5-methylphenylboronic acid 97%
3-Ethoxy-2-fluoro-5-methylphenylboronic acid
Precaución: for Research Use Only


Formula: C₉H₁₂BFO₃
MW: 197.999 g/mol
MDL Number: MFCD19981525
CAS Number: 2096333-49-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty