Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Ethyl-1H-imidazole-5-carbaldehyde hydrochloride 97%

Supplier: AMBEED, INC
Warning

A356391-1G A356391-250MG
77142-232EA 124.63 USD InStock
77142-232 77142-234
1-Ethyl-1H-imidazole-5-carbaldehyde hydrochloride 97%
1-Ethyl-1H-imidazole-5-carbaldehyde hydrochloride
Precaución: for Research Use Only


Formula: C₆H₈N₂O·ClH
MW: 160.601 g/mol
MDL Number: MFCD07186428
CAS Number: 1914148-58-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty