Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 8-amino-1-methyl-4,5-dihydro-1H-pyrazolo[4,3-h]quinazoline-3-carboxylate 97%

Supplier: AMBEED, INC
Warning

A359908-100MG A359908-1G A359908-250MG
77142-280EA 51.51 USD InStock
77142-280 77142-282 77142-284
Ethyl 8-amino-1-methyl-4,5-dihydro-1H-pyrazolo[4,3-h]quinazoline-3-carboxylate 97%
Ethyl 8-amino-1-methyl-4,5-dihydro-1H-pyrazolo[4,3-h]quinazoline-3-carboxylate
Precaución: for Research Use Only


Formula: C₁₃H₁₅N₅O₂
MW: 273.29 g/mol
MDL Number: MFCD20526553
CAS Number: 802534-49-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty