Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 1H-indol-2-ylcarbamate 95%

Supplier: AMBEED, INC
Warning

A361219-100MG A361219-1G A361219-250MG
77142-316EA 14.42 USD InStock
77142-316 77142-318 77142-320
tert-Butyl 1H-indol-2-ylcarbamate 95%
tert-Butyl 1H-indol-2-ylcarbamate
Precaución: for Research Use Only


Formula: C₁₃H₁₆N₂O₂
MW: 232.27 g/mol
MDL Number: MFCD16629876
CAS Number: 167954-49-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty