Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Amlodipine 99%

Supplier: AMBEED, INC
Danger

A367713-100G A367713-10G A367713-25G A367713-25MG A367713-5G A367713-100MG A367713-500G
77142-402EA 111.24 USD InStock
77142-402 77142-404 77142-406 77142-408 77142-410 77686-035 77686-036
Amlodipine 99%
Amlodipine
Precaución: for Research Use Only


Formula: C₂₀H₂₅ClN₂O₅
MW: 408.879 g/mol
MDL Number: MFCD00864687
CAS Number: 88150-42-9
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty