Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cediranib 98%

Supplier: AMBEED, INC
Warning

A368855-100MG A368855-10MG A368855-1MG A368855-5MG
77142-420EA 131.84 USD InStock
77142-420 77142-422 77142-424 77142-426
Cediranib 98%
Cediranib
Precaución: for Research Use Only


Formula: C₂₅H₂₇FN₄O₃
MW: 450.51 g/mol
MDL Number: MFCD09954115
CAS Number: 288383-20-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty