Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitro-3-(trifluoromethyl)phenol 98%

Supplier: AMBEED, INC
Danger

A370093-1G A370093-5G
77142-446EA 12.36 USD InStock
77142-446 77142-448
4-Nitro-3-(trifluoromethyl)phenol 98%
4-Nitro-3-(trifluoromethyl)phenol
Precaución: for Research Use Only


Formula: C₇H₄F₃NO₃
MDL Number: MFCD00014714
CAS Number: 88-30-2
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty