Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-tert-Butyl 2-hydroxypropanoate 97%

Supplier: AMBEED, INC
Warning

A371608-100MG A371608-1G A371608-250MG A371608-5G
77142-484EA 56.65 USD InStock
77142-484 77142-486 77142-488 77142-490
(R)-tert-Butyl 2-hydroxypropanoate 97%
(R)-tert-Butyl 2-hydroxypropanoate
Precaución: for Research Use Only


Formula: C₇H₁₄O₃
MDL Number: MFCD00191884
CAS Number: 68166-83-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty