Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-benzothiazolinone 98%

Supplier: AMBEED, INC
Warning

A374862-100G A374862-1G A374862-25G A374862-5G
77142-506EA 397.59 USD InStock
77142-506 77142-508 77142-510 77142-512
5-Chloro-2-benzothiazolinone 98%
5-Chloro-2-benzothiazolinone
Precaución: for Research Use Only


Formula: C₇H₄ClNOS
MDL Number: MFCD00060125
CAS Number: 20600-44-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty