Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl fluoromalonate 98%

Supplier: AMBEED, INC
Danger

A375540-10G A375540-25G A375540-5G
77142-528EA 6.18 USD InStock
77142-528 77142-530 77686-049
Diethyl fluoromalonate 98%
Diethyl fluoromalonate
Precaución: for Research Use Only


Formula: C₇H₁₁FO₄
MW: 178.16 g/mol
MDL Number: MFCD00009139
CAS Number: 685-88-1
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty