Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetradecyl acrylate 95% stabilised

Supplier: AMBEED, INC
Warning

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A384939-100G A384939-25G A384939-5G
77142-636EA 164.8 USD InStock
77142-636 77142-638 77142-640
Tetradecyl acrylate 95% stabilised
Tetradecyl acrylate
Precaución: for Research Use Only


Formula: C₁₇H₃₂O₂
MDL Number: MFCD00673244
CAS Number: 21643-42-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty