Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Cyano-4-[[(dodecylthio)carbonothioyl]thio]pentanoic acid 95%

Supplier: AMBEED, INC
Warning

A385088-1G A385088-25G A385088-5G A385088-250MG
77142-642EA 37.08 USD InStock
77142-642 77142-644 77142-646 77686-070
4-Cyano-4-[[(dodecylthio)carbonothioyl]thio]pentanoic acid 95%
4-Cyano-4-[[(dodecylthio)carbonothioyl]thio]pentanoic acid
Precaución: for Research Use Only


Formula: C₁₉H₃₃NO₂S₃
MDL Number: MFCD11040288
CAS Number: 870196-80-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty