Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-nitrobenzyl alcohol 97%

Supplier: AMBEED, INC
Danger

A395135-100MG A395135-1G A395135-250MG
77142-810EA 18.54 USD InStock
77142-810 77142-812 77142-814
2-Amino-5-nitrobenzyl alcohol 97%
2-Amino-5-nitrobenzyl alcohol
Precaución: for Research Use Only


Formula: C₇H₈N₂O₃
MDL Number: MFCD09834918
CAS Number: 77242-30-9
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty