Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethoxy)aniline 98%

Supplier: AMBEED, INC
Danger

A395838-10G A395838-25G
77142-824EA 5.15 USD InStock
77142-824 77142-826
4-(Trifluoromethoxy)aniline 98%
4-(Trifluoromethoxy)aniline
Precaución: for Research Use Only


Formula: C₇H₆F₃NO
MW: 177.13
Boiling Pt: 73
Density: 1.31
MDL Number: MFCD00041314
CAS Number: 461-82-5
UN: 2922
ADR: 8,6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty