Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-(1-Oxoisoindolin-2-yl)phenyl)butanoic acid 97%

Supplier: AMBEED, INC
Warning

A397716-1MG A397716-25MG A397716-5MG A397716-100MG
77142-864EA 6.18 USD InStock
77142-864 77142-866 77142-868 77686-102
2-(4-(1-Oxoisoindolin-2-yl)phenyl)butanoic acid 97%
2-(4-(1-Oxoisoindolin-2-yl)phenyl)butanoic acid
Precaución: for Research Use Only


Formula: C₁₈H₁₇NO₃
MDL Number: MFCD00572250
CAS Number: 63610-08-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty