Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-N-(3-bromo-4-fluorophenyl)-N'-hydroxy-1,2,5-oxadiazole-3-carboximidamide 98+%

Supplier: AMBEED, INC
Danger

A401416-10MG A401416-1MG A401416-5MG
77142-912EA 11.33 USD InStock
77142-912 77142-914 77142-916
4-Amino-N-(3-bromo-4-fluorophenyl)-N'-hydroxy-1,2,5-oxadiazole-3-carboximidamide 98+%
4-Amino-N-(3-bromo-4-fluorophenyl)-N'-hydroxy-1,2,5-oxadiazole-3-carboximidamide
Precaución: for Research Use Only


Formula: C₉H₇BrFN₅O₂
MW: 316.09 g/mol
MDL Number: MFCD28399146
CAS Number: 914638-30-5
UN: 1325
ADR: 4.1 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty