Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 2,5-dihydroxybenzoate 98%

Supplier: AMBEED, INC
Warning

A405290-10G A405290-25G A405290-5G
77142-956EA 29.86 USD InStock
77142-956 77142-958 77142-960 77142-962
Sodium 2,5-dihydroxybenzoate 98%
Sodium 2,5-dihydroxybenzoate
Precaución: for Research Use Only


Formula: C₇H₅NaO₄
MDL Number: MFCD00016504
CAS Number: 4955-90-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty