Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Hydroxylpropyl)-β-cyclodextrine 99% 19.6-26.3% (-OCH2CHOHCH3)

Supplier: AMBEED, INC
Warning

A405837-100G A405837-10G A405837-25G A405837-500G
77142-974EA 42.23 USD InStock
77142-974 77142-976 77142-978 77142-980
(2-Hydroxylpropyl)-β-cyclodextrine 99% 19.6-26.3% (-OCH2CHOHCH3)
(2-Hydroxylpropyl)-β-cyclodextrine
Precaución: for Research Use Only


Formula: C₆₃H₁₁₂O₄₂
MDL Number: MFCD16621721
CAS Number: 128446-35-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty