Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Diiodonaphthalene 97%

Supplier: AMBEED, INC
Warning

A408537-100MG A408537-1G A408537-250MG
77143-016EA 45.32 USD InStock
77143-016 77143-018 77143-020
2,6-Diiodonaphthalene 97%
2,6-Diiodonaphthalene
Precaución: for Research Use Only


Formula: C₁₀H₆I₂
MDL Number: MFCD17012279
CAS Number: 36316-88-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty