Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-hydroxybenzyl alcohol 98%

Supplier: AMBEED, INC
Warning

A435060-100G A435060-10G A435060-1G A435060-25G A435060-5G
77143-304EA 213.21 USD InStock
77143-304 77143-306 77143-308 77143-310 77143-312
5-Bromo-2-hydroxybenzyl alcohol 98%
5-Bromo-2-hydroxybenzyl alcohol
Precaución: for Research Use Only


Formula: C₇H₇BrO₂
MDL Number: MFCD00004618
CAS Number: 2316-64-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty